missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ GN (Gram-Negative) Broth, 8 mL
Click to view available options
Quantity:
8 mL
Unit Size:
Pack of 10
Pack of 100
Description
Requires
Ancillary culture media, reagents, quality control organisms and laboratory equipment
Inoculate broth as soon as possible after specimens arrive at laboratory.
Specifications
Specifications
| Certifications/Compliance | CE, BS12, CCAM, CMPH2, COMPF, MCM9 |
| Color | Light Beige |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | GN Broth |
| Formulation | Pancreatic digest of Casein 10g/L, Peptic digest of Animal Tissue 10g/L, Dextrose 1g/L, D-Mannitol 2g/L, Sodium Citrate 5g/L, Sodium Desoxycholate 0.5g/L, Dipotassium Phosphate 4g/L, Monopotassium Phosphate 1.5g/L, Sodium Chloride 5g/L |
| For Use With (Application) | Selective enrichment of Salmonella and Shigella |
| No. per Pack | 10 |
| Packaging Type | Tube |
| Show More |
For in vitro diagnostic use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction