missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Soybean Casein Digest Agar
Click to view available options
Quantity:
2.3 kg
11.3 kg
Media Type:
Soy Agar
Unit Size:
11350g
Each
Description
Applications:
Isolation of non-fastidious and fastidious
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, EP, EPA, ISO, JP, SMD, SMWW, USDA, USP |
| Content And Storage | 25°C |
| Form | Dehydrated |
| Media Type | Soy Agar |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 15g/L, Papaic digest of Soybean 5g/L, Sodium Chloride 5g/L, Agar 15g/L |
| For Use With (Application) | Isolation of non-fastidious and fastidious |
| Packaging Type | Pail |
| Quantity | 2.3 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction