missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Dehydrated Culture Media: EMB Agar, Modified, Holt-Harris and Teague

Product Code. 11768113
Click to view available options
:
EMB Agar, Modified (Holt-Harris and Teague); 500g
This item is not returnable. View return policy

Product Code. 11768113

Brand: BD 211215

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Slightly selective and differential medium for the isolation, cultivation and differentiation of gram-negative enteric bacilli from both clinical and nonclinical specimens

  • Formulation: Pancreatic digest of Gelatin 10g/L, Lactose 5g/L, Sucrose 5g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 65mg/L, Agar 13.5g/L

Certifications

CE, AOAC

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, AOAC
Content And Storage 15°C to 30°C
Form Dehydrated
Media Type EMB Agar, Modified
Product Type Agar
Formulation Pancreatic digest of Gelatin 10g/L, Lactose 5g/L, Sucrose 5g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 65mg/L, Agar 13.5g/L
For Use With (Application) Isolation, cultivation and differentiation of gram negative enteric Bacilli from both clinical and nonclinical specimen
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.