Learn More
BD BBL™ Dehydrated Culture Media: Egg Yolk Tellurite Solution
Click to view available options
:
Egg Yolk Tellurite Solution; BBL; 100mL
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | 2°C to 8°C |
| Additive | Egg Yolk Tellurite Solution |
| Form | Liquid |
| For Use With (Application) | Dehydrated culture media |
| Packaging Type | Bottle |
| Quantity | 100 mL |