missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Bacto™ Dehydrated Culture Media: Tryptic Soy Broth without Dextrose
Description
Cultivation of fastidious and non-fastidious microorganisms from variety of clinical and nonclinical specimens
Specifications
Specifications
| Certifications/Compliance | BAM |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Tryptic Soy Broth |
| Product Type | Microbiology Media |
| Formulation | Pancreatic digest of Casein 17g/L, Enzymatic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L |
| For Use With (Application) | Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?