missing translation for 'onlineSavingsMsg'
Learn More

BD Apoptosis II Sampler Kit

Product Code. 15876279
Click to view available options
:
1 kit
This item is not returnable. View return policy

Product Code. 15876279

Brand: BD 612743

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Caspase-2/ICH-1L, Caspase-3/CPP32, Caspase-7/MCH-3, DFF45, FADD, Fas/CD95/APO-1, PARP, RIP, TRADD

Western Blot

Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Sample Volume 10 μg
Storage Buffer Aqueous Buffered solution containing BSA, Glycerol and ≤0.09% Sodium Azide
Content And Storage -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.