missing translation for 'onlineSavingsMsg'
Learn More

BD-1047 dihydrobromide, MedChemExpress

Product Code. p-200095881
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487248

Brand: Medchem Express HY16996A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BD-1047 (dihydrobromide) is a selective functional antagonist of sigma-1 receptor, shows antipsychotic activity in animal models predictive of efficacy in schizophrenia.

BD-1047 (dihydrobromide) is a selective functional antagonist of sigma-1 receptor, shows antipsychotic activity in animal models predictive of efficacy in schizophrenia.

Specifications

Chemical Name or Material BD-1047 dihydrobromide
Physical Form Solid
CAS 138356-21-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C13H22Br2Cl2N2
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : 25 mg/mL (57.20 mM; Need ultrasonic) ∣H2O : 25 mg/mL (57.20 mM; Need ultrasonic)
SMILES CN(C)CCN(CCC1=CC=C(Cl)C(Cl)=C1)C.[H]Br.[H]Br
Molecular Weight (g/mol) 437.04
Formula Weight 437.04
Percent Purity 98.14%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.