missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BCLAF1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585656

Product Code. 16382585

  • 520.00 EUR / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution. Predicted reactivity: Mouse (94%), Rat (94%), Pig (98%), Bovine (98%). Positive Control: 293T, A431, HeLa, HepG2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

BCLAF1 is an intracellular, death promoting transcriptional repressor of survival genes that interacts with several members of the BCL2 family of proteins. It shows homology to the basic leucine zipper (bzip) and MYB DNA binding domains.From recent research it can be concluded that in case of apoptotic cells this protein localizes to dot-like structures throughout the nucleus, and redistributes to a zone near the nuclear envelope. When over expressed this protein induces apoptosis and can be suppressed by co-expression of BCL2 proteins. Also known as BTF, it triggers a proapoptotic signal, potentially helpful to overcome the resistance to STI571.BCLAF1 is found to express mostly in skeletal muscles.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BCLAF1
Polyclonal
Unconjugated
BCLAF1
2610102K23Rik; 2700025J07Rik; 2810454G14Rik; 5730534O06Rik; Aa2-041; AI450190; AW556225; BCL2 associated transcription factor 1; bcl-2-associated transcription factor 1; BCL2-associated transcription factor 1; Bcl-2-associated transcription factor 1-like protein; BCLAF1; BCLAF1 and THRAP3 family member 1; bK211L9.1; BP-1; BTF; Btf-s; KIAA0164; mKIAA0164
Rabbit
Antigen affinity chromatography
RUO
9774
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry, Western Blot
0.25 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Q9NYF8
BCLAF1
Recombinant protein encompassing a sequence within the C-terminus region of human BTF. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.