missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCL-3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112367
This item is not returnable.
View return policy
Description
BCL-3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
B cell Lymphoma 3 is a member of the IkB subfamily of inhibitors. This subfamily is part of the NF-kB transcription factor protein family and it is suggested that a balance in the concentration of various NF-kB family members regulates apoptosis and survival of activated T cells. Bcl-3 has been shown to have versatile functions such as cytoplasmic activation of p50 homodimers, translocation to the nucleus, and modulation of the transcriptional machinery in the nucleus. Bcl-3 activity often relies on several nuclear interacting proteins such as Tip60, Jab1, Bard1, and Pirin.
Specifications
| BCL-3 | |
| Polyclonal | |
| Unconjugated | |
| Bcl3 | |
| AI528691; B cell leukemia/lymphoma 3; B-cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3 protein homolog; B-cell lymphoma 3-encoded protein; BCL3; BCL-3; Bcl-3 protein; BCL4; chronic lymphatic leukemia protein; D19S37; Proto-oncogene BCL3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 12051, 602 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20749, Q9Z2F6 | |
| Bcl3 | |
| Recombinant Human B-cell lymphoma 3 protein (362-454AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction