Learn More
BB-K 6 N-(Benzyloxy) Carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | BB-K 6 N-(Benzyloxy) Carbamate |
| Analyte or Component Names | BB-K 6 N-(Benzyloxy) Carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C30 H49 N5 O15 |
| InChI Formula | InChI=1S/C30H49N5O15/c31-13-8-14(32)26(24(43)25(13)49-28-21(40)18(33)19(38)17(10-36)48-28)50-29-23(42)22(41)20(39)16(47-29)9-35-27(44)15(37)6-7-34-30(45)46-11-12-4-2-1-3-5-12/h1-5,13-26,28-29,36-43H,6-11,31-33H2,(H,34,45)(H,35,44)/t13-,14+,15+,16-,17-,18+,19-,20-,21-,22+,23-,24-,25+,26-,28-,29-/m1/s1 |
| Solution Type | Neat |
| SMILES | N[C@@H]1C[C@H](N)[C@@H](O[C@H]2O[C@H](CNC(=O)[C@@H](O)CCNC(=O)OCc3ccccc3)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O[C@H]4O[C@H](CO)[C@@H](O)[C@H](N)[C@H]4O |
| IUPAC Name | benzyl N-[(3S)-4-[[(2R,3S,4S,5R,6R)-6-[(1R,2R,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxy-3,4,5-trihydroxyoxan-2-yl]methylamino]-3-hydroxy-4-oxobutyl]carbamate |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 719.735 |
| Show More |