Learn More
BAZ1B Monoclonal Antibody (OTI3B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA812820
Encompass_Preferred
 View more versions of this product
Product Code. 17577434
  • 540.87 EUR / 100µL
Description
BAZ1B is a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
Specifications
Specifications
BAZ1B
Monoclonal
Unconjugated
BAZ1B
WBSCR10, WBSCR9, WSTF
Mouse
Affinity Chromatography
RUO
9031
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI3B8
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UIG0
BAZ1B
Human recombit protein fragment corresponding to amino acids 1-260 of human BAZ1B produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions