Learn More
BAZ1B Monoclonal Antibody (OTI3B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812820
This item is not returnable.
View return policy
Description
BAZ1B is a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.Specifications
| BAZ1B | |
| Monoclonal | |
| Unconjugated | |
| BAZ1B | |
| WBSCR10, WBSCR9, WSTF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9031 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3B8 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UIG0 | |
| BAZ1B | |
| Human recombit protein fragment corresponding to amino acids 1-260 of human BAZ1B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |