missing translation for 'onlineSavingsMsg'
Learn More

BAY1217389, MedChemExpress

Product Code. 16411597
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16411597

Brand: Medchem Express HY12859

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BAY 1217389 is a potent, and selective inhibitor of the monopolar spindle 1 (MPS1) kinase with an IC50 value less than 10 nM.

BAY 1217389 is a potent, and selective inhibitor of the monopolar spindle 1 (MPS1) kinase with an IC50 value less than 10 nM.

Specifications

Chemical Name or Material BAY1217389
CAS 1554458-53-5
Color White
Purity Grade Notes Research
Molecular Formula C27H24F5N5O3
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (178.09 mM)
SMILES O=C(NC1CC1)C2=CC=C(C3=CN=C4C(NCCC(F)(F)F)=CC(OC5=CC=C(OC)C(F)=C5F)=NN43)C=C2C
Molecular Weight (g/mol) 561.5
Formula Weight 561.5
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.