Learn More
BAY-545, MedChemExpress
Product Code. 16490827
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16490827
Brand: Medchem Express HY111767
MedChemExpress BAY-545 is a potent and selective A2B adenosine receptor antagonist, with an IC50 of 59 nM. BAY-545 also exhibits IC50s of 66, 400, 280 nM for human, mouse, rat A2B adenosine receptor in cells, respectively, and a Ki of 97 nM for human A2B adenosine receptor, with more selectivity over A1, A2A, and A3 adenosine receptor.
BAY-545 is a potent and selective A2B adenosine receptor antagonist, with an IC50 of 59 nM. BAY-545 also exhibits IC50s of 66, 400, 280 nM for human, mouse, rat A2B adenosine receptor in cells, respectively, and a Ki of 97 nM for human A2B adenosine receptor, with more selectivity over A1, A2A, and A3 adenosine receptor.
Specifications
Chemical Name or Material BAY-545
Physical Form Solid
CAS 1699717-32-2
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H22F3N3O4S
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 150 mg/mL (346.06 mM)
SMILES CCN1C(N(CCC(F)(F)F)C2=C(C(C)=C(C(N3CCC(O)CC3)=O)S2)C1=O)=O
Molecular Weight (g/mol) 433.45
Formula Weight 433.45
Percent Purity 95.88%
Grade Research