Learn More
Bax inhibitor 1 Antibody (20F565), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP2-24912SS
 View more versions of this product
Product Code. 18174734
  • 236.00 EUR / 25µg
Description
Bax inhibitor 1 Monoclonal specifically detects Bax inhibitor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Bax inhibitor 1
Monoclonal
0.5 mg/mL
Western Blot 1-5 ug/ml, Immunohistochemistry-Paraffin 0.5-5 ug/ml
BAXI1, BI1, BI-1Transmembrane BAX inhibitor motif-containing protein 6, TEGTbax inhibitor 1, testis enhanced gene transcript, Testis-enhanced gene transcript protein, transmembrane BAX inhibitor motif containing 6
Mouse
Protein G purified
RUO
Primary
This antibody is reactive to Human, Mouse, Rat.
Store at -20C. Avoid freeze-thaw cycles.
IgG2b κ
Western Blot, Immunohistochemistry (Paraffin)
20F565
Unconjugated
P55061
TMBIM6
A synthetic peptide made to the N-terminal portion of the human Bax inhibitor 1 protein (between amino acids 1-40) UniProt P55061.
0.025 mg
Apoptosis
7009
Human, Mouse, Rat
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions