missing translation for 'onlineSavingsMsg'
Learn More

Bavachinin, MedChemExpress

Product Code. p-200097679
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481348 5 mg 5mg
16458908 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481348 Supplier Medchem Express Supplier No. HYN0234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bavachinin(7-O-Methylbavachin) is a natural compound isolated from the Chinese herb Fructus Psoraleae;has potent anti-angiogenic activity.

Bavachinin(7-O-Methylbavachin) is a natural compound isolated from the Chinese herb Fructus Psoraleae

Specifications

Chemical Name or Material Bavachinin
CAS 19879-30-2
Color Light Nude
Purity Grade Notes Research
Molecular Formula C21H22O4
Quantity 5 mg
Synonym 7-O-Methylbavachin Bavachinin A
Solubility Information DMSO : 125 mg/mL (369.39 mM; Need ultrasonic)
SMILES O=C1C[C@@H](C2=CC=C(O)C=C2)OC3=CC(OC)=C(C/C=C(C)\C)C=C13
Molecular Weight (g/mol) 338.4
Formula Weight 338.4
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.