Learn More
Testo™ Batteries
Use this battery with a variety of products. Testo™ Batteries are compatible with Testo data loggers.
42.00 EUR - 62.45 EUR
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10002772
|
Testo™
0515 0177 |
For Testo: 175-T1, 175-T2, 177 |
62.45 EUR
Each |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10192492
|
Testo™
0515 0175 |
For Testo: 175-T3, 175-H1, 175-H2, 175-S1, 175-S2 |
42.00 EUR
Each |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- 3.6V
- 0.8Ah
- ½AA
Specifications
Battery |