Learn More
SCHOTT AG Lighting and Imaging™ Base Stand
Product Code. 11368330 Shop All Schott Products
Click to view available options
For Use With (Equipment):
Articulated Arm (158345) or Mounting Adapter (120250)
Unit Size:
Each
Product Code. 11368330
Brand: SCHOTT AG Lighting and Imaging™ 158340
Only null left
Add to basket
Add to basket
Base Stand to use with articulated arm (158345) or mounting bracket (120250).
  • Includes 3 x M6 thread.
  • Mechanical accessory designed for easy handling.
  • For use with articulating arm (fix flexible light guides or Lightlines) or mounting adapter 120250 for the standalone version with EasyLED Spotlight Plus or KL300LED.
Specifications
Type Base Stand
For Use With (Equipment) Articulated Arm (158345) or Mounting Adapter (120250)