Learn More
Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables
Maintain the performance of your Thermo Scientific™ Barnstead™ Classic Still with replacement consumables such as UV lamps and air filters.
103.00 EUR - 763.00 EUR
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
14885701
|
Thermo Scientific™
25001-DB |
Ventguard Filter Element |
103.00 EUR
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
14835731
|
Thermo Scientific™
H3120 |
replacement filter for H3111 |
346.00 EUR
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
14875721
|
Thermo Scientific™
H3130 |
Replacement Water Seal for H3111 |
763.00 EUR
Each |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
Description
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter.
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter.