Learn More
Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables
Shop All Thermo Scientific ProductsDescription
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter.
Maintain the performance of your still with Thermo Scientific™ Barnstead™ Classic Stills Replacement Consumables, including UV lamp and Thermo Scientific™ Ventgard™ filter element (the consumable portion of the Ventgard filter assembly). Prevent air from entering the reservoir via the condenser atmospheric vent and protect stored water against airborne particulates, organics and CO2 with the Ventgard air filter.
Specifications
Specifications
| For Use With (Equipment) | Barnstead Classic Stills |
| Type | Replacement Filter |
| Features | Protects stored water against airborne particulates and organics. |