Learn More
Masterflex™ BarbLock™ Ultra-Secure Retainer
Description
- BarbLock Ultra-Secure Retainer, 3/8 in. for 3/8 x 5/8 in. tubing
- Easy to install, three-piece locking mechanism ensures the tubing and hose barb won't disengage
- Safe to use with pressures up to 140psi or upper pressure limit of tubing
- An excellent alternative to securing tubing and hose barbs with cable ties
- No more crimps or tube distortion
- The BarbLock assembly consists of three pieces: the sleeve, collet and barbed fittings
- To install the fitting, slide the sleeve and collet onto the tubing and then insert the barbed fitting into the tubing
- Prelock the sleeve and collet by pressing them together until you hear a click
- Slide the sleeve and collet to the edge of the barbed fitting and lock with the BarbLock tool
- Once assembled, the BarbLock assembly can only be unlocked with the removal tool
Specifications
Specifications
| Type | Ultra-Secure Retainer |
| Diameter (English) Outer | 43/64 in. |
| Diameter (English) Inner | 0.375 in. |
| Description | BarbLock Ultra-Secure Retainer, 3/8 in. for 3/8 x 5/8 in. tubing |
| Material | Polypropylene |