missing translation for 'onlineSavingsMsg'
Learn More

BAP1 Antibody (BAP1/9795R), Novus Biologicals Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-45084

Product Code. 30448906

  • 593.00 EUR / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BAP1 Monoclonal antibody specifically detects BAP1 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BAP1
Monoclonal
1 mg/mL
Immunohistochemistry : 1-2ug/mL
BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Recombinant fragment (around aa400-729) of human BAP1 protein (Uniprot: Q92560). Exact sequence is proprietary.
100 μg
Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
8314
Store at -20 to -84°C. Avoid freeze-thaw cycles.
Purified
Immunohistochemistry
BAP1/9795R
Unconjugated
10 mM PBS
Rabbit
Protein A or G purified
RUO
Primary
Human
Antibody
IgG κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.