Learn More
Lenz Laborglasinstrumente™ Ball Members, Inch Range
Click to view available options
Overall Length (Metric):
120 mm
150 mm
Unit Size:
Each
Description
- Acc. to DIN 12264 and in addition to standard
- Made of DURAN™ tubing
Specifications
Specifications
| Certifications/Compliance | DIN 12264 |
| Material | Borosilicate Glass |
| Overall Length (Metric) | 150 mm |
| Packaging Quantity | 1 |