Learn More
Balaglitazone, MedChemExpress
Product Code. 16486978
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16486978
Brand: Medchem Express HY16086
MedChemExpress Balaglitazone is a selective partial PPARγ agonist with an EC50 of 1.351 μM for human PPARγ.
Balaglitazone is a selective partial PPARγ agonist with an EC50 of 1.351 μM for human PPARγ.
Specifications
Chemical Name or Material Balaglitazone
CAS 199113-98-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H17N3O4S
Quantity 10 mg
Synonym DRF 2593 NN 2344
Solubility Information DMSO : 100 mg/mL (252.89 mM; Need ultrasonic)
SMILES O=C(N1)SC(CC2=CC=C(OCC(N3C)=NC4=C(C=CC=C4)C3=O)C=C2)C1=O
Molecular Weight (g/mol) 395.43
Formula Weight 395.43
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism