Learn More
Gibco™ Bacto™ Proteose Peptone No. 2
Product Code. 16269771
Click to view available options
Quantity:
500 g
10 kg
Unit Size:
10kg
500g
Product Code. 16269771
Brand: Gibco™ 212120
Enzymatic digest of protein
Used in preparing microbiological culture media.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance LUO
Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type Bacterial Media
Format Bottle
For Use With (Application) Formulation of culture media for the cultivation of heterotrophic microorganisms
Product Line Bacto
Quantity 500 g
Source Animal Origin