missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bacteriophage M13 Recombinant Mouse Monoclonal Antibody (MM05), HRP, Invitrogen™
GREENER_CHOICE

Product Code. 30019461
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30019461 100 μg 100µg
30019315 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30019461 Supplier Invitrogen™ Supplier No. MA552324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Tail virion protein G7P (Bacteriophage M13) may initiate with G9P the virion concomitant assembly-budding process, by interacting with the packaging signal of the viral genome. The assembly-budding takes place at the host inner membrane. In turn, G7P and G9P are present at the end of the filamentous virion that emerges first from the bacterial host. Anti-Bacteriophage M13 Antibody is useful for researchers interested in HCP, Host Cell Membranes, Transmembrane Viruses, and E. coli bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bacteriophage M13
Applications ELISA
Classification Recombinant Monoclonal
Clone MM05
Concentration 1.18 mg/mL
Conjugate HRP
Formulation PBS with no preservative
Gene Alias Enterobacteria phage M13
Host Species Mouse
Immunogen M13 Bacteriophage.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.