Learn More
Brady™ B-7610 Paper Thermal Transfer Labels, 25 mm Core
Thermal transfer printable paper for low cost general labeling applications.
65.90 EUR - 317.00 EUR
Specifications
Autoclavable | Not Autoclavable |
---|---|
Adhesive Type | Permanent Acrylic |
Color | White |
Finish | Matte |
Material | Paper (B-7610) |
Product Code | Brand | Height (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
11598764
|
Brady™
800851 |
10 mm | 30 mm |
140.00 EUR
1 box |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
11558774
|
Brady™
800860 |
50 mm | 80 mm |
65.90 EUR
1 box |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
11548764
|
Brady™
800843 |
55.9 mm | 104 mm |
154.00 EUR
1 box |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
11588764
|
Brady™
800849 |
80 mm | 108 mm |
191.00 EUR
1 box |
Estimated Shipment: 20-05-2024
Log in to see stock. |
||||
11558764
|
Brady™
800844 |
106 mm | 108 mm |
317.00 EUR
1 box |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
Description
- 25 mm core
- Permanent acrylic adhesive
- Matte finish
Specifications
Not Autoclavable | |
White | |
Paper (B-7610) | |
Non-sterile | |
BBP™11, BBP™12, BradyPrinter i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers | |
Rectangle | |
0.081 mm |
Permanent Acrylic | |
Matte | |
Thermal Transfer | |
Hazard Communication Identification | |
1 | |
-40°C to +50°C |