Learn More
Brady™ B-7610 Paper Thermal Transfer Labels, 25 mm Core
Thermal transfer printable paper for low cost general labeling applications.
Brand: Brady™ 800851
Additional Details : Weight : 1.80000kg
Description
- 25 mm core
- Permanent acrylic adhesive
- Matte finish
Specifications
Not Autoclavable | |
White | |
Paper (B-7610) | |
Non-sterile | |
BBP™11, BBP™12, BradyPrinter i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers | |
18,000 Labels | |
-40°C to +50°C | |
30 mm |
Permanent Acrylic | |
Matte | |
Thermal Transfer | |
Hazard Communication Identification | |
1 | |
Rectangle | |
0.081 mm | |
10 mm |