missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azurocidin/CAP37/HBP Antibody (B-K37) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14596JF525
This item is not returnable.
View return policy
Description
Azurocidin/CAP37/HBP Monoclonal antibody specifically detects Azurocidin/CAP37/HBP in Human samples. It is validated for ELISA
Specifications
| Azurocidin/CAP37/HBP | |
| Monoclonal | |
| Janelia Fluor 525 | |
| AZAMP, AZU, azurocidin, azurocidin 1, CAP37Heparin-binding protein, cationic antimicrobial protein 37, HBPCationic antimicrobial protein CAP37, HUMAZUR, NAZC, neutrophil azurocidin | |
| Recombinant human Azurocidin/Cap37/HBP | |
| 0.1 mL | |
| Apoptosis | |
| 566 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA | |
| B-K37 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction