Learn More
Synergy Healthcare™ Azo™ Flat Pouches
Click to view available options
Dimensions (L x W):
250 x 75mm
150 x 100mm
210 x 150mm
250 x 100mm
300 x 100mm
320 x 150mm
420 x 200mm
No. per Case:
1200
2000
2400
2500
3600
Unit Size:
Pack of 1000
Pack of 1200
Pack of 2000
Pack of 2400
Pack of 2500
Pack of 3600
Description
- Made from specially treated paper, less fibres released when pouch is opened and reducing the risk of contamination
- New film technology increases impact strength of pouch film up to 10 times, virtually eliminates the shattering of film when opening
- Indicators are placed between the 2 layers which make-up the pouch film, prevents offsetting and leakage of ink
- Meet EN ISO 11607 part 1 and EN 868-4 standards
Specifications
Specifications
| Content And Storage | Cool dry place |
| Autoclavable | Autoclavable |
| Certifications/Compliance | EN868-5, ISO11607-1, 93/42/EEC, ISO13485, ISO9001 |
| Closure Type | Heat Seal |
| Color | Blue |
| Disposable | Single-use |
| Material | Polyethylene |
| Sterility | Non-sterile |
| Thickness (Metric) | 135 μm |
| No. per Case | 1200 |
| Show More |