Learn More
Azlon™ Spring Loaded Clamp
Description
For use with burettes. Azlon™ Burette Clamps contain rubber grips to prevent slipping.
- Spring loaded polypropylene clamps that are corrosion resistant
- Allow clear view of graduations along the full length of the burette
- Rubber grips prevent slipping
- An interchangeable compression cone on the central boss provides a firm grip on rods 11mm to 14mm in diameter
Specifications
Specifications
| Type | Burette Clamp (Double) |
| Material | Polypropylene |
| Includes | Rubber grip, interchangeable compression cone |
| For Use With (Equipment) | Burette |
| Color | White |