Learn More
Azlon™ Gas Delivery Tube
Product Code. p-8002910
Click to view available options
Overall Length (Metric):
205 mm
290 mm
335 mm
475 mm
Product Type:
Bung Top Gas Delivery Tube
Double Bend Gas Delivery Tube
Gas Delivery Tube
Side Arm Gas Delivery Tube
Unit Size:
Pack of 5
Product Code. 12673012
Brand: Azlon™ DT002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Polypropylene construction. Azlon™ Gas Delivery Tube are ideal substitutes for fragile glass alternatives.
  • Pre-bent rigid polypropylene delivery tubes
  • Unbreakable in normal use
  • Suitable for many types of gas collection experiments in schools and universities
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Diameter (English) Outer 0.23 in.
Overall Length (English) 11.41 in.
Overall Length (Metric) 290 mm
For Use With (Application) Many types of gas collection experiments in schools and universities
Max. Operating Temperature (Metric) 135°C
Product Type Bung Top Gas Delivery Tube