missing translation for 'onlineSavingsMsg'
Learn More

AZD8797, MedChemExpress

Product Code. 16456346
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16456346

Brand: Medchem Express HY13848

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD8797 (KAND567) is an allosteric non-competitive and orally active antagonist of the human CX3CR1 receptor; antagonizes CX3CR1 and CXCR2 with Kis of 3.9 and 2800 nM, respectively.

AZD8797 (KAND567) is an allosteric non-competitive and orally active antagonist of the human CX3CR1 receptor

Specifications

Chemical Name or Material AZD8797
Physical Form Solid
CAS 911715-90-7
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C19H25N5OS2
For Use With (Application) COVID-19-immunoregulation
Synonym KAND567
Solubility Information DMSO : ≥ 150 mg/mL (371.69 mM)
SMILES CC(C)C[C@@H](NC1=NC(S[C@@H](C)C2=CC=CC=C2)=NC3=C1SC(N)=N3)CO
Molecular Weight (g/mol) 403.56
Formula Weight 403.56
Percent Purity 98.65%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.