missing translation for 'onlineSavingsMsg'
Learn More

AZD8330, MedChemExpress

Product Code. p-200096852
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484958

Brand: Medchem Express HY12058

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD8330 (ARRY-424704) is a potent, uncompetitive MEK1/MEK2 inhibitor, with an IC50 of 7 nM.

AZD8330 (ARRY-424704) is a potent, uncompetitive MEK1/MEK2 inhibitor, with an IC50 of 7 nM.

Specifications

Chemical Name or Material AZD8330
CAS 869357-68-6
Color Pink
Purity Grade Notes Research
Molecular Formula C16H17FIN3O4
Quantity 10 mg
Synonym ARRY-424704 ARRY-704
Solubility Information DMSO : ≥ 100 mg/mL (216.81 mM)
SMILES IC1=CC(F)=C(C=C1)NC(N2C)=C(C=C(C2=O)C)C(NOCCO)=O
Molecular Weight (g/mol) 461.23
Formula Weight 461.23
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.