missing translation for 'onlineSavingsMsg'
Learn More

AZD2906, MedChemExpress

Product Code. p-200096687
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16483236 25 mg 25mg
16465586 100 mg 100mg
16457527 5 mg 5mg
16424788 10 mg 10mg
16411057 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16483236

Brand: Medchem Express HY113854

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD2906 is a selective glucocorticoid receptor (GR) agonist, increases micronucleated immature erythrocytes in the bone marrow of rats. AZD2906 shows IC50s of 2.2, 0.3, 41.6 and 7.5 nM at GR in human, rat PBMC and human, rat whole blood, respectively.

AZD2906 is a selective glucocorticoid receptor (GR) agonist, increases micronucleated immature erythrocytes in the bone marrow of rats. AZD2906 shows IC50s of 2.2, 0.3, 41.6 and 7.5 nM at GR in human, rat PBMC and human, rat whole blood, respectively.

Specifications

Chemical Name or Material AZD2906
CAS 1034148-15-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H25FN4O3
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (271.44 mM; Need ultrasonic)
SMILES FC1=CC=C(N2N=CC3=CC(O[C@@H]([C@@H](NC(C4CC4)=O)C)C5=CC=C(N=C5)OC)=CC=C23)C=C1
Molecular Weight (g/mol) 460.5
Formula Weight 460.5
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.