Learn More
AZD-8055, MedChemExpress
Product Code. 16440986
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Product Code. 16440986
Brand: Medchem Express HY10422
MedChemExpress AZD-8055 is a potent, selective, and orally bioavailable ATP-competitive mTOR kinase inhibitor with an IC50 of 0.8 nM. AZD-8055 inhibits both mTORC1 and mTORC2.
AZD-8055 is a potent, selective, and orally bioavailable ATP-competitive mTOR kinase inhibitor with an IC50 of 0.8 nM. AZD-8055 inhibits both mTORC1 and mTORC2.
Specifications
Chemical Name or Material AZD-8055
CAS 1009298-09-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H31N5O4
Quantity 200 mg
Solubility Information DMSO : 33.33 mg/mL (71.59 mM; Need ultrasonic)
SMILES OCC1=CC(C2=NC3=NC(N4CCOC[C@@H]4C)=NC(N5[C@@H](C)COCC5)=C3C=C2)=CC=C1OC
Molecular Weight (g/mol) 465.54
Formula Weight 465.54
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease