missing translation for 'onlineSavingsMsg'
Learn More

AZD 6482, MedChemExpress

Product Code. p-200097012
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16496567

Brand: Medchem Express HY10344

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD 6482 (KIN-193) is a potent and selective p110β inhibitor with an IC50 of 0.69 nM.

AZD 6482 (KIN-193) is a potent and selective p110β inhibitor with an IC50 of 0.69 nM.

Specifications

Chemical Name or Material AZD 6482
Physical Form Solid
CAS 1173900-33-8
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H24N4O4
For Use With (Application) Cancer-Kinase/protease
Synonym KIN-193
Solubility Information DMSO : ≥ 100 mg/mL (244.83 mM)
SMILES O=C1N2C(C([C@H](NC3=CC=CC=C3C(O)=O)C)=CC(C)=C2)=NC(N4CCOCC4)=C1
Molecular Weight (g/mol) 408.45
Formula Weight 408.45
Percent Purity 98.85%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.