missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZ7550 Mesylate, MedChemExpress
Description
AZ7550 Mesylate is an active metabolite of AZD9291 and inhibits the activity of IGF1R with an IC50 of 1.6 μM.
Specifications
Specifications
| Chemical Name or Material | AZ7550 Mesylate |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C30H43N7O11S3 |
| Quantity | 5 mg |
| Synonym | AZ7550 trimesylate salt |
| Solubility Information | DMSO : 25 mg/mL (32.30 mM; Need ultrasonic) |
| SMILES | CN1C=C(C2=NC(NC3=CC(NC(C=C)=O)=C(N(CCNC)C)C=C3OC)=NC=C2)C4=CC=CC=C41.O=S(O)(C)=O.O=S(O)(C)=O.O=S(O)(C)=O |
| Molecular Weight (g/mol) | 773.9 |
| Formula Weight | 773.9 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?