missing translation for 'onlineSavingsMsg'
Learn More

AZ PFKFB3 26, MedChemExpress

Product Code. 16433668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433668 10 mg 10mg
16406407 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16433668 Supplier Medchem Express Supplier No. HY101971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZ PFKFB3 26 is a potent and selective inhibitor of the metabolic kinase PFKFB3 with an IC50 of 23 nM. AZ PFKFB3 26 inhibits PFKFB1 and PFKFB2 with IC50s of 2.06 and 0.384 μM, respectively.

AZ PFKFB3 26 is a potent and selective inhibitor of the metabolic kinase PFKFB3 with an IC50 of 23 nM. AZ PFKFB3 26 inhibits PFKFB1 and PFKFB2 with IC50s of 2.06 and 0.384 μM, respectively.

Specifications

Chemical Name or Material AZ PFKFB3 26
CAS 1704740-52-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C24H26N4O2
Quantity 10 mg
Solubility Information DMSO : 5 mg/mL (12.42 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES O=C([C@H]1NCCC1)NC2=CC=C(OC3=CC4=C(N(CC(C)C)C=C4C#N)C=C3)C=C2
Molecular Weight (g/mol) 402.49
Formula Weight 402.49
Percent Purity 98.38%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.