missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZ 5104, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | N-(5-((4-(1H-Indol-3-yl)pyrimidin-2-yl)amino)-2-((2-(dimethylamino)ethyl)(methyl)amino)-4-methoxyphenyl)acrylamide |
| CAS | 1421373-98-9 |
| Molecular Formula | C27H31N7O2 |
| Quantity | 25 mg |
| Synonym | AZ 5104, N-(5-((4-(1H-Indol-3-yl)pyrimidin-2-yl)amino)-2-((2-(dimethylamino)ethyl)(methyl)amino)-4-methoxyphenyl)acrylamide |
| InChI Formula | InChI=1S/C27H31N7O2/c1-6-26(35)30-22-15-23(25(36-5)16-24(22)34(4)14-13-33(2)3)32-27-28-12-11-21(31-27)19-17-29-20-10-8-7-9-18(19)20/h6-12,15-17,29H,1,13-14H2,2-5H3,(H,30,35)(H,28,31,32) |
| SMILES | COc1cc(N(C)CCN(C)C)c(NC(=O)C=C)cc1Nc2nccc(n2)c3c[nH]c4ccccc34 |
| IUPAC Name | N-[2-[2-(dimethylamino)ethyl-methylamino]-5-[[4-(1H-indol-3-yl)pyrimidin-2-yl]amino]-4-methoxyphenyl]prop-2-enamide |
| Molecular Weight (g/mol) | 485.58 |
| Formula Weight | 485.2539 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction