missing translation for 'onlineSavingsMsg'
Learn More

AZ-5104, MedChemExpress

Product Code. 16498456
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498456 100 mg 100mg
16474487 50 mg 50mg
16441068 5 mg 5mg
16438618 10 mg 10mg
16421446 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498456 Supplier Medchem Express Supplier No. HYB0793

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZ-5104 is an active, demethylated metabolite of AZD 9291. AZ-5104 is an EGFR inhibitor with IC50s of 1, 6, 1, 25 and 7 nM for EGFRL858R/T790M, EGFRL858R, EGFRL861Q, EGFR and ErbB4, respectively.

AZ-5104 is an active, demethylated metabolite of AZD 9291. AZ-5104 is an EGFR inhibitor with IC50s of 1, 6, 1, 25 and 7 nM for EGFRL858R/T790M, EGFRL858R, EGFRL861Q, EGFR and ErbB4, respectively.

Specifications

Chemical Name or Material AZ-5104
CAS 1421373-98-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C27H31N7O2
Quantity 100 mg
Solubility Information DMSO : ≥ 28 mg/mL (57.66 mM)
SMILES C=CC(NC1=CC(NC2=NC=CC(C3=CNC4=C3C=CC=C4)=N2)=C(OC)C=C1N(CCN(C)C)C)=O
Molecular Weight (g/mol) 485.58
Formula Weight 485.58
Percent Purity 99.34%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.