Learn More
Axygen™ Tip Refill System, Empty Racks
Product Code. 11340285 Shop All Axygen Scientific Products
Change view
Click to view available options
Model:
RFL-R
RFL-R-1200
Packaging:
10 Racks/Pack; 3 Packs/Case
10 Racks/Pack; 5 Packs/Case
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Packaging unitSize
11340285 RFL-R-1200 10 Racks/Pack; 3 Packs/Case Pack of 10
11330285 RFL-R 10 Racks/Pack; 5 Packs/Case Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11340285 Supplier Axygen™ Supplier No. RFLR1200
Only null left
Add to basket
Add to basket
Easy-to-load refill racks for Axygen Tip Refill System. For use with P2/P10 Gilson-Style and 200μL Universal fit inserts.
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
  • RFL-R is suitable for both the 10 μL and 200 μL refill inserts
  • RFL-R-1200 is suitable for 1200 μL refill inserts
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Sterility Non-sterile
For Use With (Equipment) P2/P10 Gilson-Style and 200μL Universal fit inserts
Model RFL-R-1200
Packaging 10 Racks/Pack; 3 Packs/Case
Product Type Empty Refill Rack with Lid