Learn More
Axygen™ Tecan™ Evo MCA96 Compatible Nested Pipet Tips
BIOPHARMAGREENER_CHOICE
Product Code. 15277668 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
50 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Gray
Orange
Unit Size:
Pack of 3840
Product Code. 15277668
Brand: Axygen™ EV50NTRS
Only null left
Add to basket
Add to basket
768 tips for use with Tecan Evo MCA96 multi-channel arm
  • Tip format: 96 tip
  • Tip volume range: 50 μL to 200 μL
  • Tip material: Clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
  • Available options:
    - With or without aerosol-resistant filters
    - Nonsterile or sterile
  • Compatible heads: MCA96
  • Rack format: ANSI/SLAS
  • Rack material: Polypropylene
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 50 μL
Sterility Sterile
Color Orange
Tip Style Round
Packaging Nested tip rack
Quantity 40 Racks/Cs.
For Use With (Equipment) Tecan EVO MCA96
Product Line Tecan