Learn More
Axygen™ Slicone 96 Round Well Compression Flat Mat for PCR Microplates
Shop All Axygen Scientific Products
Click to view available options
Material:
Silicone
Unit Size:
Pack of 50
Description
Axygen™ Compression Mats are ideally suited to help reduce evaporation and ensure a proper seal when used in conjunction with Axygen PCR Microlates and PlateMaxTM sealing films. Made of inert 100% virgin silicone, they are capable of withstanding numerous PCR cycles.
For use with 96/384 well and PCR microplates
Specifications
Specifications
| Color | Natural |
| Material | Silicone |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| No. of Wells | 96 |
| No. per Case | 50/Cs. |
| No. per Pack | 10/Pk. |
| Product Type | Compression Mat |
| Well Shape | Round |
| Plate Compatibility | 96/384-well PCR Microplate |