missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Septa Caps for 0.5mL ABI Prism 310 Sequencer Tubes
Shop All Axygen Scientific Products
Click to view available options
For Use With (Equipment):
0.5 mL ABI Prism 310 Sequencer Tubes
Unit Size:
Pack of 500
Description
Axygen™ AxyMatsTM Individual Septa Cap is designed to be used on ABI sequencer. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.
Individual septa caps for 0.5 mL ABI Prism 310 sequencer tubes
Specifications
Specifications
| For Use With (Equipment) | 0.5 mL ABI Prism 310 Sequencer Tubes |
| Product Type | Septa Caps |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction