Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing
Shop All Axygen Scientific Products
Click to view available options
Color:
Amber
Clear
Sterility:
Non-sterile
Sterile
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Unit Size:
Pack of 4000
Pack of 500
Beschreibung
Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.
- Maximum G-force is 20,000 RCF.
- RNase-/DNase-free
- Nonpyrogenic
Tubes, with Cap, Sterile
All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.
Colored-cap versions are also available in sterile format.
Bestellinformation
Caps sold separately.
Spezifikation
Spezifikation
| Cap Style | Screw Cap |
| Certifications/Compliance | RNase, DNase, pyrogen-free |
| Capacity (Metric) | 0.5 mL |
| Color | Clear |
| Material | Polypropylene |
| No. per Pack | 500/Pk. |
| Packaging | 500 Tubes/Pk., 8 Packs/Cs. |
| Sterility | Non-sterile |
| Graduated | No |
| Closure Type | Not Included |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur