Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing
Product Code. 11384225 Shop All Axygen Scientific Products
Click to view available options
Color:
Amber
Clear
Sterility:
Non-sterile
Sterile
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Unit Size:
Pack of 4000
Pack of 500
Product Code. 11384225
Brand: Axygen™ ST200SS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Self-standing tubes
Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.
  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic
Tubes, with Cap, Sterile
All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.
Colored-cap versions are also available in sterile format.
Order Info
Caps sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw Cap
Certifications/Compliance RNase, DNase, pyrogen-free
Capacity (Metric) 2 mL
Color Clear
Material Polypropylene
No. per Pack 500/Pk.
Packaging 500 Tubes/Pk., 8 Packs/Cs.
Sterility Non-sterile
Graduated No
Closure Type Not Included
Bottom Shape Conical
Max. RCF 20,000 x g