Learn More
Axygen™ Replacement Electrodes for 7cm Gel Box
Accessory for use with 7cm horizontal gel box
104.00 EUR
Specifications
For Use With (Equipment) Gel Box, 7 cm
View More Specs
Products 2
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
15543177
View Documents Promotion Details
Axygen™
HGB7-NE
Replacement Positive Electrode
104.00 EUR
1 unit
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
to see stock.
Add to basket
15553177
View Documents Promotion Details
Axygen™
HGB7-PE
Replacement Negative Electrode
104.00 EUR
1 unit
View alternative products
Save Up to 
»
Estimated Shipment: 26-07-2024
Description
Replacement electrode
Specifications
Specifications
Gel Box, 7 cm