Learn More
Axygen™ Presterilized Conical Centrifuge Tubes
GREENER_CHOICE
Product Code. 11344205 Shop All Axygen Scientific Products
Click to view available options
Capacity (Metric):
15 mL
50 mL
Packaging:
25 Tubes and Caps/Rack, 20 Racks/Cs.
25 Tubes and Caps/Unit, 20 Units/Cs.
50 Tubes and Caps/Unit, 20 Units/Cs.
Bag
Unit Size:
Pack of 25
Pack of 500
Product Code. 11344205
Brand: Axygen™ SCT50ML25S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Graduated tubes with conical bottom
  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw Cap
Autoclavable Not Autoclavable
Capacity (Metric) 50 mL
Color Clear
Material Polypropylene
For Use With (Application) For sample identification
Includes Screw cap
Packaging 25 Tubes and Caps/Unit, 20 Units/Cs.
Sterility Sterile
Bottom Shape Conical
Max. RCF 12,000 x g