Learn More
Axygen™ Presterilized Conical Centrifuge Tubes
Shop All Axygen Scientific Products
Click to view available options
Capacity (Metric):
15 mL
50 mL
Packaging:
25 Tubes and Caps/Rack, 20 Racks/Cs.
25 Tubes and Caps/Unit, 20 Units/Cs.
50 Tubes and Caps/Unit, 20 Units/Cs.
Bag
Unit Size:
Pack of 25
Pack of 500
Description
- Rated to 12,000 RCF
- Printed graduations and writing area for sample identification
- Screw caps can be closed with one hand.
- Sterile options are also available.
- RNase-/DNase-free
- Nonpyrogenic
Specifications
Specifications
| Cap Style | Screw Cap |
| Closure Material | HDPE |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 15 mL |
| Color | Clear |
| Material | Polypropylene |
| For Use With (Application) | For sample identification |
| Includes | Screw cap |
| No. per Pack | 25/Pk. |
| Packaging | Bag |
| Show More |