Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Spécification
Specifications
| Color | Natural |
| Material | Silicone |
| Sterility | Non-sterile |
| Autoclavable | Autoclavable |
| No. of Wells | 384 |
| No. per Case | 50/Cs. |
| No. per Pack | 10/Pk. |
| Product Type | Sealing Mat |
| Well Shape | Square |
| Plate Compatibility | 384-well Deep Well Plate with Square Wells |